ContactUsLinkComponent
DNA ladder 500bp MiniOne. Supplied in ready-to-use format and consists of ten double-strand DNA fragments with sizes of 5K-4,5K-4K-3 ,5K-3K-2,5K-2K-1,5K-1K-500 base pairs. (50 loads) 1 * 500 µl
  EMBIM3145
 :  EMBITEC
DNA ladder 500bp MiniOne. Supplied in ready-to-use format and consists of ten double-strand DNA fragments with sizes of 5K-4,5K-4K-3 ,5K-3K-2,5K-2K-1,5K-1K-500 base pairs. (50 loads) 1 * 500 µl
  EMBIM3145
 :  EMBITEC
Frequently Bought Together