ContactUsLinkComponent
Gel tray platform silver grey MiniOne. Silver gray colour plastic plate with texture on well area and features for one orientation f it. 64x42mm. 1 * 10 items
Catalog # EMBIM2015
Supplier:  EMBITEC
Gel tray platform silver grey MiniOne. Silver gray colour plastic plate with texture on well area and features for one orientation f it. 64x42mm. 1 * 10 items
Catalog # EMBIM2015
Supplier:  EMBITEC
Frequently Bought Together