ContactUsLinkComponent
Mix dNTP MiniOne. A ready-to-use aqueous solution containing dATP, dCTP, dGTP and dTTP, each at a final concentration of 10mM. 1 * 1 mL
Catalog # EMBIM6203
Supplier:  EMBITEC
Mix dNTP MiniOne. A ready-to-use aqueous solution containing dATP, dCTP, dGTP and dTTP, each at a final concentration of 10mM. 1 * 1 mL
Catalog # EMBIM6203
Supplier:  EMBITEC
Frequently Bought Together