ContactUsLinkComponent
MiniLab Restriction analysis of DNA with digested controls. Includes: two restriction enzymes, enzyme reaction buffer, 5X sample loa ding dye, DNA fragment, three predigested and one undigested DNA controls, MiniOne universal DNA marker, 10 x 1,5% ag 1 * 1 KIT
Catalog # EMBIM6053
Supplier:  EMBITEC
MiniLab Restriction analysis of DNA with digested controls. Includes: two restriction enzymes, enzyme reaction buffer, 5X sample loa ding dye, DNA fragment, three predigested and one undigested DNA controls, MiniOne universal DNA marker, 10 x 1,5% ag 1 * 1 KIT
Catalog # EMBIM6053
Supplier:  EMBITEC
Frequently Bought Together