ContactUsLinkComponent
Centrifuge single speed MiniOne. Portable mini centrifuge with fixed speed 10000rpm (4800xg maximum rcf). AC 100V-240V 50-60Hz. Incl udes two interchangeable rotors for 6x1,5/2,0ml microcentrifuge tubes, 2 strips of 8 PCR tubes, tube adapters for 0, 1 * 1 items
Catalog # EMBIM2032
Supplier:  EMBITEC
Centrifuge single speed MiniOne. Portable mini centrifuge with fixed speed 10000rpm (4800xg maximum rcf). AC 100V-240V 50-60Hz. Incl udes two interchangeable rotors for 6x1,5/2,0ml microcentrifuge tubes, 2 strips of 8 PCR tubes, tube adapters for 0, 1 * 1 items
Catalog # EMBIM2032
Supplier:  EMBITEC
Frequently Bought Together