ContactUsLinkComponent
Tip micropipette 2-200µl universal fit with standardisation marks clear non-sterile 1 * 1.000 items
Catalog # EMBIM3134
Supplier:  EMBITEC
Tip micropipette 2-200µl universal fit with standardisation marks clear non-sterile 1 * 1.000 items
Catalog # EMBIM3134
Supplier:  EMBITEC
Frequently Bought Together