ContactUsLinkComponent
Shaker KM CO2 - FL 2.245,00 40,00 1.347,00 incl. basic platform and rubber mat Ideal for use in CO2 incubators Shaking motion: orbital Shakting platform: 200 x 295 mm Max. Loading: 2 kg Shakting speed: 5 - 220 rpm (in steps of 5) Stroke: 20 mm CO2 co 1 * 1 items
Catalog # BUHL6137000
Shaker KM CO2 - FL 2.245,00 40,00 1.347,00 incl. basic platform and rubber mat Ideal for use in CO2 incubators Shaking motion: orbital Shakting platform: 200 x 295 mm Max. Loading: 2 kg Shakting speed: 5 - 220 rpm (in steps of 5) Stroke: 20 mm CO2 co 1 * 1 items
Catalog # BUHL6137000
Frequently Bought Together