ContactUsLinkComponent
HEMATOXYLIN IS FREQUENTLY USED TOGETHER WITH EOSIN, NUCLEAR FAST RED, PERIODIC ACID OR ALCIAN BLUE. IT IS ALSO USED IN IMMUNOHISTOCH EMICAL PROCEDURES TO VISUALIZE THE END PRODUCT. OUR WEIGERT’S HEMATOXYLIN KIT DOES NOT CONTAIN MERCURY. 1 * 500 mL
Catalog # GENOBAQ084
Supplier:  G-Biosciences
HEMATOXYLIN IS FREQUENTLY USED TOGETHER WITH EOSIN, NUCLEAR FAST RED, PERIODIC ACID OR ALCIAN BLUE. IT IS ALSO USED IN IMMUNOHISTOCH EMICAL PROCEDURES TO VISUALIZE THE END PRODUCT. OUR WEIGERT’S HEMATOXYLIN KIT DOES NOT CONTAIN MERCURY. 1 * 500 mL
Catalog # GENOBAQ084
Supplier:  G-Biosciences
Frequently Bought Together