ContactUsLinkComponent
pZerve is a ready-to-use, sterile, cryopreservation solution that does not contain dimethyl sulfoxide (DMSO), fetal bovine serum or other animal protein. 1 * 60 mL
Catalog # 0920303M2
Dodavatel:  MP Biomedicals
pZerve is a ready-to-use, sterile, cryopreservation solution that does not contain dimethyl sulfoxide (DMSO), fetal bovine serum or other animal protein. 1 * 60 mL
Catalog # 0920303M2
Dodavatel:  MP Biomedicals
Frequently Bought Together