ContactUsLinkComponent
Mouse IgG monoclonal antibody for RPS6 detection. Tested with Tested with WB, IHC-P, ICC/IF, FCM in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTM01567
Supplier:  Boster Bio
Mouse IgG monoclonal antibody for RPS6 detection. Tested with Tested with WB, IHC-P, ICC/IF, FCM in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTM01567
Supplier:  Boster Bio
Frequently Bought Together