ContactUsLinkComponent
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects TREML1 in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
  BSBTEZ1521
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects TREML1 in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
  BSBTEZ1521
Frequently Bought Together