ContactUsLinkComponent
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects IL-7 in Human s amples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
  BSBTEZ0779
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects IL-7 in Human s amples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
  BSBTEZ0779
Frequently Bought Together