ContactUsLinkComponent
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Angiopoietin-1 in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
Catalog # BSBTEZ0559
Supplier:  Boster Bio
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Angiopoietin-1 in Human samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
Catalog # BSBTEZ0559
Supplier:  Boster Bio
Frequently Bought Together