ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Histone H1.0/H1F0 detection. Tested with WB, IHC-P, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA08821-1
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Histone H1.0/H1F0 detection. Tested with WB, IHC-P, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA08821-1
Supplier:  Boster Bio
Frequently Bought Together