ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detec tion of Human TIM-3/HAVCR2. 96wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTFEK1644
Supplier:  Boster Bio
Sandwich High Sensitivity ELISA kit for Quantitative The Fast version of Picokine ELISA kits, assay takes less than 1.5 hours. Detec tion of Human TIM-3/HAVCR2. 96wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTFEK1644
Supplier:  Boster Bio
Frequently Bought Together