ContactUsLinkComponent
Detect Mouse Mip 1 Gamma/Ccl9 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultu re supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1225
Supplier:  Boster Bio
Detect Mouse Mip 1 Gamma/Ccl9 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultu re supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK1225
Supplier:  Boster Bio
Frequently Bought Together