ContactUsLinkComponent
Detect Rat Mip 1Alpha/CCL3 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK1219
Supplier:  Boster Bio
Detect Rat Mip 1Alpha/CCL3 with <1pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK1219
Supplier:  Boster Bio
Frequently Bought Together