ContactUsLinkComponent
Detect Human Pdgf Bb/PDGFB with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0484
Supplier:  Boster Bio
Detect Human Pdgf Bb/PDGFB with <2pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTFEK0484
Supplier:  Boster Bio
Frequently Bought Together