Detect Mouse Fibronectin/FN1 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell cultu re supernates, serum and plasma(heparin, EDTA, citrate). 1 * 1 KIT
Catalog # BSBTFEK0351
Supplier: Boster Bio
Frequently Bought Together