ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Integrin alpha 3 detection. Tested with WB, IHC-P, IHC-F, ICC/IF, FCM in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA02902
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Integrin alpha 3 detection. Tested with WB, IHC-P, IHC-F, ICC/IF, FCM in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA02902
Supplier:  Boster Bio
Frequently Bought Together