ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative Detection of rat FGFR4. 142wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTEK2085
Supplier:  Boster Bio
Sandwich High Sensitivity ELISA kit for Quantitative Detection of rat FGFR4. 142wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTEK2085
Supplier:  Boster Bio
Frequently Bought Together