ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human TNFRSF21. 96wells/kit, with removable strips. 1 * 1 KIT
  BSBTEK2004
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human TNFRSF21. 96wells/kit, with removable strips. 1 * 1 KIT
  BSBTEK2004
Frequently Bought Together