ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human ANGPTL6. 96wells/kit, with removable strips. 1 * 1 KIT
Katalogové číslo BSBTEK1993
Supplier:  Boster Bio
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human ANGPTL6. 96wells/kit, with removable strips. 1 * 1 KIT
Katalogové číslo BSBTEK1993
Supplier:  Boster Bio
Frequently Bought Together