Rabbit IgG polyclonal antibody for MEK4/MAP2K4 detection. Tested with WB, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA01725-1
Supplier: Boster Bio
Rabbit IgG polyclonal antibody for MEK4/MAP2K4 detection. Tested with WB, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA01725-1
Supplier: Boster Bio
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection