ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Cortactin detection. Tested with WB, IHC-P, IHC-F, ICC, FCM, IF, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA01253-1
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Cortactin detection. Tested with WB, IHC-P, IHC-F, ICC, FCM, IF, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA01253-1
Supplier:  Boster Bio
Frequently Bought Together