ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human Omentin/intelectin-1. 96wells/kit, with removable strips. 1 * 1 KIT
  BSBTEK1849
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human Omentin/intelectin-1. 96wells/kit, with removable strips. 1 * 1 KIT
  BSBTEK1849
Frequently Bought Together