ContactUsLinkComponent
Rabbit IgG polyclonal antibody for PKC delta/PRKCD detection. Tested with WB, IHC-P, IHC-F, ICC, FCM, Direct ELISA in Human Mouse Ra t. 1 * 100 µG
Catalog # BSBTA00822-1
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for PKC delta/PRKCD detection. Tested with WB, IHC-P, IHC-F, ICC, FCM, Direct ELISA in Human Mouse Ra t. 1 * 100 µG
Catalog # BSBTA00822-1
Supplier:  Boster Bio
Frequently Bought Together