ContactUsLinkComponent
This antibody is purified from antiserum by immunoaffinity chromatography which removes essentially all goat serum proteins, except the specific antibody for rabbit IgG. 1 * 0,5 mL
Catalog # BSBTBA1142
Supplier:  Boster Bio
This antibody is purified from antiserum by immunoaffinity chromatography which removes essentially all goat serum proteins, except the specific antibody for rabbit IgG. 1 * 0,5 mL
Catalog # BSBTBA1142
Supplier:  Boster Bio
Frequently Bought Together