ContactUsLinkComponent
Avidin is a 68KD protein that extracted from egg white. It has very high affinity to biotin molecule, one million times than the com mon affinity between antigen and antibody. 1 * 0,5 mL
Catalog # BSBTBA1128
Supplier:  Boster Bio
Avidin is a 68KD protein that extracted from egg white. It has very high affinity to biotin molecule, one million times than the com mon affinity between antigen and antibody. 1 * 0,5 mL
Catalog # BSBTBA1128
Supplier:  Boster Bio
Frequently Bought Together