ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human Serpin A1. 96wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTEK1634
Supplier:  Boster Bio
Sandwich High Sensitivity ELISA kit for Quantitative Detection of human Serpin A1. 96wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTEK1634
Supplier:  Boster Bio
Frequently Bought Together