Mouse IgG monoclonal antibody for Cofilin 2/CFL2 detection. Tested with WB, IHC-P, ICC/IF, FCM in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTM04773
Supplier: Boster Bio
Mouse IgG monoclonal antibody for Cofilin 2/CFL2 detection. Tested with WB, IHC-P, ICC/IF, FCM in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTM04773
Supplier: Boster Bio
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection