ACRYLAMIDE-2,3,3 D3 1 * 10 mg
Catalog # EHERC10045301
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Melting point standards
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Heneicosanoic acid methyl ester certified 1 * 0,1 g
Supplier: EHRENSTORFER
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Benzo(g,h,i)perylene D12 10 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER