ContactUsLinkComponent
VOLUMETRIC PIPETTE 17 ML, CLASS AS, BLUE GRADUATED, TOLERANCE: ±0,030 ML, COLOR-CODE: -SPECIAL SIZE VOLUMETRIC PIPETTES, TOLERANCES ON THE BASIS OF DIN EN ISO 648 1 * 6 items
Catalog # WITG3.535.170.BL
Supplier:  witeg Labortechnik
VOLUMETRIC PIPETTE 17 ML, CLASS AS, BLUE GRADUATED, TOLERANCE: ±0,030 ML, COLOR-CODE: -SPECIAL SIZE VOLUMETRIC PIPETTES, TOLERANCES ON THE BASIS OF DIN EN ISO 648 1 * 6 items
Catalog # WITG3.535.170.BL
Supplier:  witeg Labortechnik
Frequently Bought Together