ContactUsLinkComponent
VOLUMETRIC PIPETTES, 4,5 ML, CLASS AS, WITH ONE MARK, CONFORMITY CERTIFIED, WAITING TIME 5 S., AMBER DIFFICO, PACK = 12 PCS. 1 * 12 items
Catalog # WITG3.535.045.BL
Supplier:  witeg Labortechnik
VOLUMETRIC PIPETTES, 4,5 ML, CLASS AS, WITH ONE MARK, CONFORMITY CERTIFIED, WAITING TIME 5 S., AMBER DIFFICO, PACK = 12 PCS. 1 * 12 items
Catalog # WITG3.535.045.BL
Supplier:  witeg Labortechnik
Frequently Bought Together