YELLOWISH GREEN (YG) STOCK REFERENCE 1 * 100 mL
Catalog # REGECPSRSYG01
Supplier: REAGECON
Frequently Bought Together
Ethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol 99.5+%
Supplier: Thermo Fisher Scientific
1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals