ContactUsLinkComponent
Microslide pink 90 ground edges alu unimark made of soda lime glass of 3. hydrolytic class in compliance with din iso 8037/1 , dimensions: approx. 76 x 26 mm, thickness approx. 1 mm. 1 * 2.500 items
Catalog # 630-3075
Supplier:  MARIENFELD
Microslide pink 90 ground edges alu unimark made of soda lime glass of 3. hydrolytic class in compliance with din iso 8037/1 , dimensions: approx. 76 x 26 mm, thickness approx. 1 mm. 1 * 2.500 items
Catalog # 630-3075
Supplier:  MARIENFELD
Frequently Bought Together