ContactUsLinkComponent
Flask volumetric pyrex, class A, amber coloured, 25 ml, ukas certified 1 * 1 items
Catalog # BIBB3152/08AUKAS
Supplier:  SciLabware
Flask volumetric pyrex, class A, amber coloured, 25 ml, ukas certified 1 * 1 items
Catalog # BIBB3152/08AUKAS
Supplier:  SciLabware
Frequently Bought Together