Water estimator quickfit with stopcock , capacity 10ml, socket size 24/29, cone size 24/29 1 * 1 items
BIBBWE6/33/10T
Water estimator quickfit with stopcock , capacity 10ml, socket size 24/29, cone size 24/29 1 * 1 items
BIBBWE6/33/10T
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection