ContactUsLinkComponent
BactoView™ Live Red: Red dye to stain live and dead bacteria ?Ex/?Em (with DNA) = 572/675 nm Detect in the PE-Cy®5 channel for flow cytometry, and the Cy®3 channel for microscopy 1 * 100 µl
Catalog # 40101.
Supplier:  Biotium
BactoView™ Live Red: Red dye to stain live and dead bacteria ?Ex/?Em (with DNA) = 572/675 nm Detect in the PE-Cy®5 channel for flow cytometry, and the Cy®3 channel for microscopy 1 * 100 µl
Catalog # 40101.
Supplier:  Biotium
Frequently Bought Together