ContactUsLinkComponent
Pycnometer to Bingham 25ml calibrated NS 7.5 glass stopper, DIN 12807 Boro 3.3 accuracy 3 decimals Weight less than 30g 1 * 1 items
Catalog # NEUB0312-50-025
Pycnometer to Bingham 25ml calibrated NS 7.5 glass stopper, DIN 12807 Boro 3.3 accuracy 3 decimals Weight less than 30g 1 * 1 items
Catalog # NEUB0312-50-025
Frequently Bought Together