ContactUsLinkComponent
ChromegaChiral(TM) Chiral CC2, 5 Micron HPLC Column, 5 mum particle size, L x I.D. 15 cm x 20 mm 1 * 1 items
Catalog # SIALES5654
Supplier:  Merck
ChromegaChiral(TM) Chiral CC2, 5 Micron HPLC Column, 5 mum particle size, L x I.D. 15 cm x 20 mm 1 * 1 items
Catalog # SIALES5654
Supplier:  Merck
Frequently Bought Together