ContactUsLinkComponent
Flash-chromatography assembly with threaded joints, capacity 200 mL, With fritted disc (porosity C), Joint: ST/NS 29/32, column O.D. x L 25 mm x 584 mm 1 * 1 items
Catalog # SIALZ416711-1EA
Supplier:  Merck
Flash-chromatography assembly with threaded joints, capacity 200 mL, With fritted disc (porosity C), Joint: ST/NS 29/32, column O.D. x L 25 mm x 584 mm 1 * 1 items
Catalog # SIALZ416711-1EA
Supplier:  Merck
Frequently Bought Together