ContactUsLinkComponent
Flash-chromatography assembly with threaded joints, capacity 100 mL, With fritted disc (porosity C), Joint: ST/NS 29/32 1 * 1 items
Catalog # SIALZ416703-1EA
Dodavatel:  Merck
Flash-chromatography assembly with threaded joints, capacity 100 mL, With fritted disc (porosity C), Joint: ST/NS 29/32 1 * 1 items
Catalog # SIALZ416703-1EA
Dodavatel:  Merck
Frequently Bought Together