ContactUsLinkComponent
TLC Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, C18-silica gel 60 matrix, binder, Polymeric, fluorescent indicator 1 * 1 Each
Catalog # SIALZ293032-1PAK
Supplier:  Merck
TLC Glass Plates from Merck KGaA, Darmstadt, Germany, L x W 20 cm x 20 cm, C18-silica gel 60 matrix, binder, Polymeric, fluorescent indicator 1 * 1 Each
Catalog # SIALZ293032-1PAK
Supplier:  Merck
Frequently Bought Together