ContactUsLinkComponent
ChromegaChiral(TM) Chiral CC3, 5 Micron HPLC Column, 5 mum particle size, L x I.D. 10 cm x 3.0 mm 1 * 1 items
Catalog # SIALES5681
Supplier:  Merck
ChromegaChiral(TM) Chiral CC3, 5 Micron HPLC Column, 5 mum particle size, L x I.D. 10 cm x 3.0 mm 1 * 1 items
Catalog # SIALES5681
Supplier:  Merck
Frequently Bought Together