ContactUsLinkComponent
Proteomix(R) SAX-NP10 Strong Anion Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 4 mm, 10 mum particle size, stainless steel 1 * 1 items
Catalog # SIALZ777202
Supplier:  Merck
Proteomix(R) SAX-NP10 Strong Anion Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 4 mm, 10 mum particle size, stainless steel 1 * 1 items
Catalog # SIALZ777202
Supplier:  Merck
Frequently Bought Together