ContactUsLinkComponent
Proteomix(R) WCX-NP1.7 Weak Cation Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 4 mm, 1.7 mum particle size, stainless steel 1 * 1 items
Catalog # SIALZ777178
Supplier:  Merck
Proteomix(R) WCX-NP1.7 Weak Cation Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 4 mm, 1.7 mum particle size, stainless steel 1 * 1 items
Catalog # SIALZ777178
Supplier:  Merck
Frequently Bought Together