ContactUsLinkComponent
Screw-cap for flash-chromatography assembly with threaded joints, Joint: ST/NS 45/40, Joint: ST/NS 45/50 fits both joints 1 * 4 items
Catalog # SIALZ147567-4EA
Supplier:  Merck
Screw-cap for flash-chromatography assembly with threaded joints, Joint: ST/NS 45/40, Joint: ST/NS 45/50 fits both joints 1 * 4 items
Catalog # SIALZ147567-4EA
Supplier:  Merck
Frequently Bought Together