ContactUsLinkComponent
Columns for flash-chromatography assemblies with ball joints, Standard column, capacity 100 mL, Joint: SJ 28/12 1 * 1 items
Katalogové číslo SIALZ106976-1EA
Dodavatel:  Merck
Columns for flash-chromatography assemblies with ball joints, Standard column, capacity 100 mL, Joint: SJ 28/12 1 * 1 items
Katalogové číslo SIALZ106976-1EA
Dodavatel:  Merck
Frequently Bought Together