ContactUsLinkComponent
Proteomix(R) SCX-NP1.7 Strong Cation Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 2 mm, 1.7 mum particle size, stainless ste el 1 * 1 items
Catalog # SIALZ777141
Supplier:  Merck
Proteomix(R) SCX-NP1.7 Strong Cation Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 2 mm, 1.7 mum particle size, stainless ste el 1 * 1 items
Catalog # SIALZ777141
Supplier:  Merck
Frequently Bought Together